Racks for freezer
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5400009 5400003 5400035 5400036 5400037 5400038 5400040 5400041 5400042 5400002 5400005 5400022 5400024 5400006 5400032 5400033 5400034
RATI5400009EA 166 EUR
RATI5400009 RATI5400003 RATI5400035 RATI5400036 RATI5400037 RATI5400038 RATI5400040 RATI5400041 RATI5400042 RATI5400002 RATI5400005 RATI5400022 RATI5400024 RATI5400006 RATI5400032 RATI5400033 RATI5400034
Racks for freezer
Rekken Rekken voor cryogene dozen
For the storing of cryoboxes in freezer cabinets and chests.
  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning
Leveringsinformatie: Cryo racks are supplied with shelf sizes for any standard cryoboxes.