Screw caps for internal thread sample tubes
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T500NOS T500ROS T500YOS T500BOS T500GOS T500WOS T500LOS T500OOS
212-9335EA 237 EUR
212-9335 SIMPT500ROS SIMPT500YOS SIMPT500BOS SIMPT500GOS SIMPT500WOS SIMPT500LOS SIMPT500OOS
Screw caps for internal thread sample tubes
Sluitingen Reageerbuissluitingen
These polypropylene screw caps are to be used with T500 series internal thread sample tubes.
  • Made of polypropylene
  • Cap with silicone washer for leakproof seal
  • Long skirt and super-fast thread design for easy seal
  • Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
These silicone washer caps will guarantee a positive leakproof seal at allMore Product Information