Heptylbenzene ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N-heptylbenzene, 1-Phenylheptane
A12969.14
A12969.22
A12969.36
A12969.14EA
53.1
EUR
A12969.14
A12969.22
A12969.36
Heptylbenzene ≥98%
Heptylbenzene
Purity: 98%
CAS No.: 1078-71-3
Molecular Formula: C13H20
Formula Weight: 176.30
Flash Point: 95°(203°F)
Refractive Index: 1.4850
Melting Point: -48°
Boiling Point: 230-233°
Beilstein Reference: 1905782
UN No.: 3082
MDL No.: MFCD00009545
Formule:
C₁₃H₂₀ Kookpunt: 230…233 °C Smeltpunt: –48 °C Dichtheid: 0,860 Vlampunt: 95 °C (203 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009545 CAS nummer: 1078-71-3 EINECS: 214-084-3 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...