Kranen, éénweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286455404 286455601 286456006 286456409 286457002 286457208
SCOT286455404EA 770 EUR
SCOT286455404 SCOT286455601 SCOT286456006 SCOT286456409 SCOT286457002 SCOT286457208
Kranen, éénweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For PTFE keys
Straight bore.
Order Now
Specificaties
Environmentally Preferable