Kranen, éénweg, 1:5, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286453009 286453806 286453206 286453403 286454005 286454202
SCOT286453009EA 1120 EUR
SCOT286453009 SCOT286453806 SCOT286453206 SCOT286453403 SCOT286454005 SCOT286454202
Kranen, éénweg, 1:5, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, with PTFE key.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.