Kranen, tweeweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286424808 286425804
SCOT286424808EA 357 EUR
SCOT286424808 SCOT286425804
Kranen, tweeweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance