Kranen, tweeweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286424208 286425007 286425607 286426003 286422807
SCOT286424208EA 545 EUR
SCOT286424208 SCOT286425007 SCOT286425607 SCOT286426003 SCOT286422807
Kranen, tweeweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE key.
  • Very good chemical resistance
  • Very good temperature resistance
Two oblique bores.