Kranen, tweeweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286423006 286425204
SCOT286423006EA 232 EUR
SCOT286423006 SCOT286425204
Kranen, tweeweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance