Kranen, éénweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286442003 286442809 286443008 286443805 286444201 286444407 286446005 286446605 286446802 286447001 286445403
SCOT286442003EA 105 EUR
SCOT286442003 SCOT286442809 SCOT286443008 SCOT286443805 SCOT286444201 SCOT286444407 SCOT286446005 SCOT286446605 SCOT286446802 SCOT286447001 SCOT286445403
Kranen, éénweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
Straight bore.