Kranen, éénweg, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286438005 286439001 286439207 286439601 286439807 286440002 286440405 286440602 286441007 286441401 286441607
SCOT286438005EA 165 EUR
SCOT286438005 SCOT286439001 SCOT286439207 SCOT286439601 SCOT286439807 SCOT286440002 SCOT286440405 SCOT286440602 SCOT286441007 SCOT286441401 SCOT286441607
Kranen, éénweg, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3,3, clear, polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
  • For glass manipulators
Ungrounded, without key.