Stopcocks, Two-way
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8614-0007 8614-0009
BURK8614-0007EA 5.5 EUR
BURK8614-0007 BURK8614-0009
Stopcocks, Two-way
Afsluitkranen
HDPE, natural.
  • Resistant to acids and chemicals
  • Pressure resistant up to 0,3 bar
  • Good temperature resistance
Simple reflux nozzle with two-sided hose nozzle.