Orion™ RDO™ Dissolved Oxygen Probes
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
087010MD 087020MD
664-0154EA 1790 EUR
664-0154 664-0155
Orion™ RDO™ Dissolved Oxygen Probes
Elektroden Elektroden voor opgeloste zuurstof
These RDO (Rugged Dissolved Oxygen) probes use optical DO technology for problem-free measurement.
  • Measure dissolved oxygen levels without the difficulties of membrane replacement
  • Once-a-year maintenance with a simple cap replacement
  • Different cable lengths available
  • IP 68-rated, waterproof
Can be used with Orion Star meters with RDO input.
Leveringsinformatie: Supplied with optical cap, stainless steel sensor guard and calibration sleeve.