Tubing, PVC
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8875-0608 8801-0710 8801-0608
BURK8875-0608EA 44.7 EUR
BURK8875-0608 BURK8801-0710 BURK8801-0608
Tubing, PVC
Slangen Transfer Tubing
Tubing made of soft PVC, cadmium-free, with high resistance to chemicals, high flexibility and permanent crystal clear transparency and excellent abrasion resistance.
  • For pressureless operation
  • Suitable for use with footstuffs
Operating temperature: −20 to +50 °C
Hardness (Shore durometer A): 80.
Certificaten: Foodstuffs quality according to the EC Guideline 1935/2004.