Kranen voor scheitrechters, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286432208 286432405 286432602 286432808 286433007
SCOT286432208EA 663 EUR
SCOT286432208 SCOT286432405 SCOT286432602 SCOT286432808 SCOT286433007
Kranen voor scheitrechters, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, with short inlet, with glass or PTFE key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.