Extension rods, with M10 thread
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2159 2161 2163
USBE2159EA 22.5 EUR
USBE2159 USBE2161 USBE2163
Extension rods, with M10 thread
Statiefstaven
Extension rods made from stainless steel.
  • One internal and one external thread M 10
  • Ø 12 mm
  • Size of jaw 10 mm
Order Now
Specificaties
Environmentally Preferable