Powder funnels, PP/PE
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9606-0350 9606-0240 9606-0080 9606-0100 9606-0150
BURK9606-0350EA 45.4 EUR
BURK9606-0350 BURK9606-0240 BURK9606-0080 BURK9606-0100 BURK9606-0150
Powder funnels, PP/PE
Trechters Poedertrechters
These powder funnels are designed for safe filling of containers.
  • PP/PE
9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).