Trityl chloride for synthesis
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane
M-Clarity™ quality level = MQ200
8.08348.0250 8.08348.1000
MERC8.08348.0250EA 91.2 EUR
MERC8.08348.0250 MERC8.08348.1000
Trityl chloride for synthesis
Trityl chloride
Formule: C₁₉H₁₅Cl
MW: 278.78 g/mol
Kookpunt: 374
Smeltpunt: 109…112 °C
Dichtheid: 1.25 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000813
CAS nummer: 76-83-5
EINECS: 200-986-4
UN: 1759
ADR: 8 II