Kranen voor scheitrechters, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286430207 286430404 286430601
SCOT286430207EA 585 EUR
SCOT286430207 SCOT286430404 SCOT286430601
Kranen voor scheitrechters, DURAN®
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.
Order Now
Specificaties
Environmentally Preferable