Kranen voor scheitrechters
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286430807 286431006 286431203
SCOT286430807EA 427 EUR
SCOT286430807 SCOT286431006 SCOT286431203
Kranen voor scheitrechters
Afsluitkranen
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
With short inlet. For glass manipulators.