Kranen voor scheitrechters, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286433401 286433607 286433804
SCOT286433401EA 643 EUR
SCOT286433401 SCOT286433607 SCOT286433804
Kranen voor scheitrechters, DURAN®
Afsluitkranen
DURAN®, borosilicate glass, with polished short funnel, without key.
  • Very good chemical resistance
  • High temperature resistance
For glass manipulators.