2,4-Dimethylbenzaldehyde 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
165790050 165790250 165791000
ACRO165790050EA 29.3 EUR
ACRO165790050 ACRO165790250 ACRO165791000
2,4-Dimethylbenzaldehyde 99%
2,4-Dimethylbenzaldehyde
Formule: C₉H₁₀O
MW: 134.18 g/mol
Kookpunt: 102.4…103 °C (14 mmHg)
Smeltpunt: –9 °C
Dichtheid: 0.96 g/cm³
Vlampunt: 88 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003340
CAS nummer: 15764-16-6
EINECS: 239-856-7
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5482 to 1.5502 (20°C, 589 nm)
GC ≥98.5 %