5-Methyl-2-thenoic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

AC12788DA AC12788EA AC12788.50
MAYBAC12788DAEA 0 EUR
MAYBAC12788DA MAYBAC12788EA MAYBAC12788.50
5-Methyl-2-thenoic acid
5-Methyl-2-thenoic acid
Formule: C₆H₆O₂S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005439
CAS nummer: 1918-79-2
EINECS: 217-640-3

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 136°C to 139°C
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR