5-Bromothiophene-2-carbaldehyde
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 5-Bromo-2-thiophenecarboxaldehyde
AC16665.1 AC16665.10 AC16665.50
MAYBAC16665.1EA 0 EUR
MAYBAC16665.1 MAYBAC16665.10 MAYBAC16665.50
5-Bromothiophene-2-carbaldehyde
5-Bromothiophene-2-carbaldehyde
Formule: C₅H₃BrOS
MDL Nummer: MFCD00005432
CAS nummer: 4701-17-1
EINECS: 225-176-8