Benzylammonium chloride 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
200540050
200541000
ACRO200540050EA
19
EUR
ACRO200540050
ACRO200541000
Benzylammonium chloride 99%
Benzylammonium chloride
Formule:
C₇H₁₀ClN Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00012852 CAS nummer: 3287-99-8 EINECS: 221-943-6 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 259 to 264 °C |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...