2-Aminobenzenesulphonic acid 95%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
400640250 400641000 400645000
ACRO400640250EA 21 EUR
ACRO400640250 ACRO400641000 ACRO400645000
2-Aminobenzenesulphonic acid 95%
2-Aminobenzenesulphonic acid
Formule: C₆H₇NO₃S
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007705
CAS nummer: 88-21-1
EINECS: 201-810-9
UN: 2585
ADR: 8,III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥94 %
GC ≥96.0 %