2-Aminobenzamidoxime 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H51782.03 H51782.06
H51782.03EA 93 EUR
H51782.03 H51782.06
2-Aminobenzamidoxime 97%
2-Aminobenzamidoxime

Purity: 97% CAS No.: 16348-49-5 Molecular Formula: C7H9N3O Formula Weight: 151.17 Melting Point: 71-75° MDL No.: MFCD00492723

Formule: C₇H₉N₃O
Smeltpunt: 71…75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00492723
CAS nummer: 16348-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR