4-Bromothiophene-2-carbaldehyde
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde
CC28504.1 CC28504.10 CC28504.50
MAYBCC28504.1EA 0 EUR
MAYBCC28504.1 MAYBCC28504.10 MAYBCC28504.50
4-Bromothiophene-2-carbaldehyde
4-Bromothiophene-2-carbaldehyde
Formule: C₅H₃BrOS
MDL Nummer: MFCD00005431
CAS nummer: 18791-75-8
EINECS: 242-577-3