Cyclododecanone oxime
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RH01554.250 RH01554.1
MAYBRH01554.250EA 0 EUR
MAYBRH01554.250 MAYBRH01554.1
Cyclododecanone oxime
Cyclododecanone oxime
Formule: C₁₂H₂₃NO
CAS nummer: 946-89-4