Ethanolamine hydrochloride
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.20063.0100 8.20063.0500
8.20063.0100EA 36.8 EUR
8.20063.0100 8.20063.0500
Ethanolamine hydrochloride
Ethanolamine hydrochloride
Formule: C₂H₈ClNO
MDL Nummer: MFCD00012892
CAS nummer: 2002-24-6
EINECS: 217-900-6
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 80 - 84 °C
Molar Mass 97.54 g/mol