4-Isopropoxyaniline ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
I0399-25G
I0399-5G
I0399-100G
TCIAI0399-25GEA
130
EUR
TCIAI0399-25G
TCIAI0399-5G
TCIAI0399-100G
4-Isopropoxyaniline ≥98.0% (by GC, titration analysis)
4-Isopropoxyaniline
Formule:
C₉H₁₃NO Kookpunt: 95°C |
MDL Nummer:
MFCD00059188 CAS nummer: 7664-66-6 EINECS: 231-636-9 |
|
Specification Test Results
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...