PCR racks
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6114102 6114100 6114103 6114104
RATI6114102EA 20.7 EUR
RATI6114102 RATI6114100 RATI6114103 RATI6114104
PCR racks
Rekken Tube Racks
Racks made of PP and are characterised by their very thin but uniform walls.
  • 8×12 positions, micro test plate format
  • Autoclavable
  • Available in different colors for the convenient identification of different types of samples
  • Transparent lids available in two different heights (Lids must be ordered seperately)
Work and storage rack for 0,2 ml and 0,5 ml PCR tubes, PCR strips, and PCR segments.