Cleanibags, ratiolab®
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7000260 7000270 7000250
RATI7000260EA 164 EUR
RATI7000260 RATI7000270 RATI7000250
Cleanibags, ratiolab®
Zakken Monsterzakken
All-purpose PE bags with self-adhesive strip for clinical and laboratory use.
  • Self-adhesive strip: sticks on smooth and clean surfaces (e.g. walls, work benches, beds, bedside tables)
  • Residue-free, removable
Bestelinformatie: Other sizes or models available on request, e.g. for intensive care and bags with custom-made imprint.
Verpakking: 1000 pieces per cardboard box.