Wipes for general applications, Tork
Leverancier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
130082 130083 130043
115-1608EA 80.8 EUR
115-1608 115-0634 115-1605
Wipes for general applications, Tork
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor algemeen gebruik
Tork Plus: All-round wipe for cleaning surfaces, objects and hands
Tork Industrial Heavy-Duty: Strong and bulky wipe, ideal for engineering and mechanical environments.
  • Food contact approved
  • Industrial Heavy-Duty: Fast absorption of oil and lubricants improves productivity
Colour: White or blue.
Order Now
Specificaties
Environmentally Preferable