Sample cups for analysers, ratiolab®
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5700401 5700200 5700120 5700110
RATI5700401EA 28.5 EUR
RATI5700401 RATI5700200 RATI5700120 RATI5700110
Sample cups for analysers, ratiolab®
Monsternamepotjes Monsternamepotjes voor analysatoren
Made of clear PS.
  • Good chemical resistance
  • Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.