Mercury collectors
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9789-0001 9789-0002
BURK9789-0001EA 30.5 EUR
BURK9789-0001 BURK9789-0002
Mercury collectors
Absorptiematerialen en bescherming tegen morsen
This mercury collector enables the easy collection of spilt mercury for reusal.
  • Screw cap lid with integrated foam pad to absorb mercury
  • Perforated mesh insert in the container
  • Pad releases mercury when squeezed against mesh insert by screwing lid onto the jar