Zakken, biologisch gevaarlijk materiaal
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7002100 7002150 7002200 7002300
RATI7002100EA 288 EUR
RATI7002100 RATI7002150 RATI7002200 RATI7002300
Zakken, biologisch gevaarlijk materiaal
Zakken Afvalzakken
These disposable waste bags are made from strong PP film with a broad seam at the bottom. Intended for the sterilisation and disposal of hazardous and contaminated labware.
  • Rectangle white indicator patch field turns into dark when autoclaved
  • Bags must not be completely closed while autoclaving
Available with biohazard sticker and safety imprint in English, German, French and Spanish language.