PCR plates, 96-well
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8610014 8610011
RATI8610014EA 457 EUR
RATI8610014 RATI8610011
PCR plates, 96-well
Microplaten PCR-platen
These plates conform fully to the highest requirements of PCR. They are made of polypropylene and are characterised by their very thin but uniform walls. In this way, optimal heat transfer in short cycle times is achieved. The smooth inside walls of the cavities minimise the binding of enzymes and nucleic acids.
  • Free from DNA, DNase and RNase
  • Available without or with rims (the latter to minimise cross-contamination).
  • Alphanumeric coding for optimal sample identification
  • Available in three versions for various PCR systems
Working volume: 0,2 ml