Funnels with short stem
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8840 8841 8842 8843 8844 8845
221-0284EA 29.2 EUR
221-0284 221-0285 221-0286 221-0287 221-0288 221-0289
Funnels with short stem
Trechters Filtertrechters Filtertrechters
Stainless steel ¹⁸/₁₀.
  • Good chemical resistance, extremely high thermal resistance
  • Conical air tube
  • Autoclavable
Smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye (exeption 80 mm Ø) for easy storage.