Genormaliseerde dubbele slijpstukken, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286172809 286173205 286174004 286174407 286569808 286571608
SCOT286172809EA 330 EUR
SCOT286172809 SCOT286173205 SCOT286174004 SCOT286174407 SCOT286569808 SCOT286571608
Genormaliseerde dubbele slijpstukken, DURAN®
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
DIN 12249