Immersiecirculatiethermostaten, geavanceerd, Optima™ TX150 en TXF200
Leverancier: GRANT INSTRUMENTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T-BTS TXF200 TX150
461-0779EA 203 EUR
461-0779 461-0664 461-0656
Immersiecirculatiethermostaten, geavanceerd, Optima™ TX150 en TXF200
Circulatiethermostaten Open circulatiesystemen Thermostaatregelaars
The Optima™ range of advanced thermostats can be combined with plastic baths, premium stainless steel baths or refrigeration units. The TX150 and TXF200 thermostats feature a full colour QVGA TFT screen displaying actual and set temperature and clear status icons. The QVGA TFT screen, in combination with the easy to use dial and function keys, allows rapid navigation through the intuitive menu system. Both units have RS232 and USB interfaces and can be programmed via remote PC or laptop using Grant’s versatile Labwise™ software (optional). Additional features include countdown timer, adjustable over-temperature cut-out, visual, audible and programmable high and low temperature alarms. The TX150 has the capacity to store one program with 30 segments. The TXF200 can also be programmed via the user interface and can store up to 10 programs containing 100 segments. The programming interface includes set target temperature with a choice of time to target temperature or temperature ramp speed and an additional programmable relay for on/off control of ancillary equipment or refrigeration units.
  • TX150 temperature range: Ambient 5 to 150 °C. Settable range −50 to +150 °C (operation below ambient requires accessory cooling)
  • TXF200 temperature range: Ambient 5 to 200 °C. Settable range −50 to +200 °C (operation below ambient requires accessory cooling)
  • Stability: ±0,01 °C at 70 °C
  • External PT1000 temperature probe socket
  • Offset adjustment for isothermal performance
  • Five-point user calibration of internal and external probes (two fixed points and three user defined)
Maximale pompcapaciteiten:
TX150 18 L/min, 310 mbar
TXF200 22 L/min, 530 mbar variable speed
Bestelinformatie: Optional T-clamp (included with some models) allows attachment of thermostat onto virtually any vertical sided bath or vessel.
Opgelet: *RT = Ambient.