Cytology funnels, CytoSep™ for the Wescor Cytopro® cytocentrifuge
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M967-20FW M967-10FW
720-1959EA 366 EUR
720-1959 720-1957
Cytology funnels, CytoSep™ for the Wescor Cytopro® cytocentrifuge
Trechters Cytologische trechters
These funnels will snap quickly in place, allowing the pad to align correctly with the sample tunnel. They are made with a compression ring around the sample hole in order to better control the rate of absorption, and to ensure more consistent results. Two ports, one in the sample well and one in the tunnel, provide great versatility. Samples are loaded directly through the chamber caps to prevent spilling of hazardous ones. Caps provide added safety to the operator. A large, centred cell deposit area makes screening easier and more sensitive. Double funnels with two chambers enable cellMore Product Information