Cleaning cloths, Tork Industrial
Leverancier: Essity
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
520337 520304
115-0638EA 82.3 EUR
115-0638 115-3122
Cleaning cloths, Tork Industrial
Schoonmaakdoekjes Herbruikbare reinigingsdoeken Doeken voor algemeen gebruik
These cleaning cloths made from virgin cellulose pulp, polyester fibres and PP fibres are a perfect alternative to rags or rental cloths.
  • exelCLEAN® technology – for a quicker professional cleaning result
  • Strong texture for excellent oil and water absorption
  • Meet all cleaning needs due to their suitability with most solvents and detergents
  • Tork Easy Handling™ for easier carrying, opening and disposing of packaging
  • Silicone-free: Ensures that there are no residues on a surface
  • Suitable for different dispensing systems
Their softness and flexibility allows cleaning tight spaces and intricate components without scratching the surface, yet they are tough enough to mop up oil, grease and dirt.
Colour: Grey