Stirrer shafts, propeller type, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C378-04 C378-06 C378-08 C378-12 C378-14 C378-16 C378-18 C378-20 C378-22
BOHLC378-04EA 260 EUR
BOHLC378-04 BOHLC378-06 BOHLC378-08 441-2915 BOHLC378-14 441-2930 BOHLC378-18 BOHLC378-20 BOHLC378-22
Stirrer shafts, propeller type, BOLA
Roerders Roerassen
Stirrer shafts made of PTFE-jacketed stainless steel and the propeller completely made of PTFE.
  • With three 45° angled round or angular blades
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C