Bottles, narrow neck, square, PET, with tamper-proof closure
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0370-0250 0370-0500 0370-0125 0370-1000
BURK0370-0250EA 2.2 EUR
BURK0370-0250 BURK0370-0500 BURK0370-0125 BURK0370-1000
Bottles, narrow neck, square, PET, with tamper-proof closure
Flessen Kunststof flessen
PET, crystal-clear.
  • Cap absolutely tight closing
  • Suitable for use with foodstuffs
  • Sterile
  • Tamper-proof closure
  • Moulded-in raised graduation