Microtube racks
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8200003 8200004 8200023 8200024 8200007
RATI8200003EA 616 EUR
RATI8200003 RATI8200004 RATI8200023 RATI8200024 RATI8200007
Microtube racks
Rekken Tube Racks
Racks, made of PP, translucent, for micro tubes, feature a highly practical system for processing small volumes and a stable robust system having a dust tight lid and also available as empty racks or equipped with coded or non-coded.
  • Side edges labeled with a barcode
  • Stackable to use with stacker instruments
  • Sloping edge helps the positioning of the refill unit
  • Temperature resistant to −80 °C