Screw caps and pouring rings for laboratory bottles
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9180003430 9180003432 9180003417 9180001588
201-1821EA 3.1 EUR
201-1821 201-1825 201-1826 KAVA9180001588
Screw caps and pouring rings for laboratory bottles
Sluitingen Flesdoppen
Made of PP or PBT
  • For DIN GL threads
  • Heat resistant up to 140 °C (PP) and up to 180 °C (PBT)
  • Autoclavable