Cyclohexylbenzene 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Phenylcyclohexane
111291000 111295000
ACRO111291000EA 0 EUR
ACRO111291000 ACRO111295000
Cyclohexylbenzene 98%
Cyclohexylbenzene
Formule: C₁₂H₁₆
Kookpunt: 239 °C (1013 hPa)
Smeltpunt: 5 °C
Dichtheid: 0,95 g/cm³
Vlampunt: 98 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001451
CAS nummer: 827-52-1
EINECS: 212-572-0
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20°C, 589 nm)
GC ≥97.5 %