Cyclohexylbenzene ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Phenylcyclohexane
A16320.22
A16320.36
A16320.22EA
62
EUR
A16320.22
A16320.36
Cyclohexylbenzene ≥97%
Cyclohexylbenzene
Purity: 97+%
CAS No.: 827-52-1
Molecular Formula: C12H16
Formula Weight: 160.26
Flash Point: 98°(208°F)
Refractive Index: 1.5260
Odour: Mild
Melting Point: 5-6°
Boiling Point: 239-240°
Beilstein Reference: 1906803
UN No.: 3082
MDL No.: MFCD00001451
Formule:
C₁₂H₁₆ Kookpunt: 239…240 °C Smeltpunt: 5…6 °C Dichtheid: 0,945 Vlampunt: 98 °C (208 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001451 CAS nummer: 827-52-1 EINECS: 212-572-0 UN: 3082 ADR: 9,III |