4-Phenylbenzoic acid 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
106280250 106281000 106285000
ACRO106280250EA 48 EUR
ACRO106280250 ACRO106281000 ACRO106285000
4-Phenylbenzoic acid 99%
4-Phenylbenzoic acid
Formule: C₁₃H₁₀O₂
MW: 198.22 g/mol
Kookpunt: 373
Smeltpunt: 223…228 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002553
CAS nummer: 92-92-2
EINECS: 202-203-1
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 223 to 228 °C
HPLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %