6-Quinolinecarboxylic acid
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
CC04501.250
CC04501DA
MAYBCC04501.250EA
0
EUR
MAYBCC04501.250
MAYBCC04501DA
6-Quinolinecarboxylic acid
6-Quinolinecarboxylic acid
Formule:
C₁₀H₇NO₂ MW: 173.17 g/mol Kookpunt: 345 Smeltpunt: 294…295 °C |
MDL Nummer:
MFCD00047613 CAS nummer: 10349-57-2 EINECS: 233-761-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...